missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7072011
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001204 100 g
6001206 250 g
6001205 1 kg
6001207 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6001204 Supplier Honeywell Inc Supplier No. 31252100G
Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥97.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Quantity 100 g
Linear Formula KCN
UN Number 1680
Beilstein 3593645
Formula Weight 65.12g/mol
Percent Purity ≥97%
Grade ACS/puriss. p.a.
Chemical Name or Material Potassium Cyanide, Reag. Ph. Eur.
Hazard Statement P260-P264-P273-P280-P284-P301 + P310
Precautionary Statement H290-H300 + H310 + H330-H370-H372-H410
EINECSNumber 205-792-3
RTECSNumber TS8750000