missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, ACS Reagent, Reagents

Catalog No. p-200021187
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337854 125 g
01337853 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337854 Supplier Labchem Inc Supplier No. C1282000125D2
Only null left

Potassium Cyanide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.12
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
IUPAC Name potassium iminomethanide
SMILES [K+].[C-]#N

Specifications

Boiling Point 1625°C
Melting Point 634.5°C
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.52
Chemical Name or Material Potassium Cyanide, ACS Reagent