missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, ACS Reagent, Reagents

Catalog No. 01337854
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337854 125 g
01337853 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337854 Supplier Labchem Inc Supplier No. C1282000125D2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Cyanide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.12
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
IUPAC Name potassium iminomethanide
SMILES [K+].[C-]#N

Specifications

Boiling Point 1625°C
Melting Point 634.5°C
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.52
Chemical Name or Material Potassium Cyanide, ACS Reagent