missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, ACS, 96.0% min, Thermo Scientific Chemicals

Catalog No. AA1213618
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1213618 50 g
AA1213630 250 g
AA12136A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1213618 Supplier Thermo Scientific Chemicals Supplier No. 01213618

KCN is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von Richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Boiling Point 1625°C
Melting Point 634°C
Physical Form Granules
Quantity 50 g
Assay Percent Range 96.0% min.
Beilstein 4652394
Solubility Information Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol.
Formula Weight 65.12
Grade ACS Reagent
Sensitivity Hygroscopic
Density 1.52
Chemical Name or Material Potassium Cyanide, For analysis ACS
Show More Show Less
Hazard Category H290-H300+H310+H330-H372
Precautionary Statement P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P390-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 205-792-3
RTECSNumber TS8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only