missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium cyanide, 97+%, extra pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Spécifications
Spécifications
| Boiling Point | 1625.0°C |
| Melting Point | 634.0°C |
| Color | Colorless |
| Physical Form | Liquid |
| Quantity | 500 g |
| Assay Percent Range | 97% min. (Argentometry) |
| Linear Formula | CKN |
| Merck Index | 15,7747 |
| Solubility Information | Solubility in water: 400g/L (20°C). Other solubilities: ethanol 4.5g/L (19.5°C) |
| SMILES | [C-]#N.[K+] |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May be corrosive to metals. Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure if inhaled. Very toxic to aquatic life with |
| Precautionary Statement | GHS P Statement Absorb spillage to prevent material damage. Rinse mouth. IF ON SKIN: Gently wash with plenty of soap and water. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at r |
| EINECSNumber | 205-792-3 |
RUO – Research Use Only