Learn More
Potassium cyanate, 97%
CAS: 590-28-3 | CKNO | 81.12 g/mol
$72.01 - $669.05
Chemical Identifiers
| CAS | 590-28-3 |
|---|---|
| Molecular Formula | CKNO |
| Molecular Weight (g/mol) | 81.12 |
| MDL Number | MFCD00011411 |
| InChI Key | GKKCIDNWFBPDBW-UHFFFAOYSA-M |
| Synonym | potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 |
| PubChem CID | 11378442 |
| ChEBI | CHEBI:38904 |
| SMILES | [K+].[O-]C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA3335622
|
Thermo Scientific Chemicals
03335622 |
100 g |
Each for $72.01
|
|
|||||
|
AA3335636
|
Thermo Scientific Chemicals
03335636 |
500 g |
Each for $188.89
|
|
|||||
|
AA33356A3
|
Thermo Scientific Chemicals
033356A3 |
2 kg |
Each for $669.05
|
|
|||||
Description
Potassium cyanate is used in gold mining, organic synthesis and electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 590-28-3 | |
| 81.12 | |
| GKKCIDNWFBPDBW-UHFFFAOYSA-M | |
| 11378442 | |
| [K+].[O-]C#N |
| CKNO | |
| MFCD00011411 | |
| potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 | |
| CHEBI:38904 |
Specifications
| 700°C to 900°C (decomposition) | |
| White | |
| 100 g | |
| MFCD00011411 | |
| potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688 | |
| GKKCIDNWFBPDBW-UHFFFAOYSA-M | |
| potassium;cyanate | |
| 11378442 | |
| 81.12 | |
| Odorless | |
| Potassium cyanate |
| 590-28-3 | |
| Crystalline | |
| CKNO | |
| 14,7625 | |
| Soluble in water; Slightly soluble in alcohol | |
| [K+].[O-]C#N | |
| 81.12 | |
| CHEBI:38904 | |
| 97% | |
| 2.056 g/mL |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 209-676-3
RTECSNumber : GS6825000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only