missing translation for 'onlineSavingsMsg'
Learn More

Potassium citrate tribasic monohydrate, 99%, extra pure

Catalog No. p-4380105
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC244210010 1 kg
AC244210050 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC244210010 Supplier Thermo Scientific Chemicals Supplier No. 244210010
Only null left

CAS: 6100-05-6 | C6H5K3O7·H2O | 324.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H5K3O7·H2O
Molecular Weight (g/mol) 324.42
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
PubChem CID 2735208
ChEBI CHEBI:64746
IUPAC Name tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]

Specifications

Melting Point 180.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula KO2CCH2C(OH)(CO2K)CH2CO2K·H2O
Quantity 1 kg
Merck Index 15,7744
Solubility Information Solubility in water: 640g/L (25°C).
Formula Weight 324.42
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Potassium citrate tribasic monohydrate
Show More Show Less

RUO – Research Use Only