missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate), Reagents

Catalog No. p-200020915
Change view
Click to view available options
Quantity:
125 g
2.5 kg
500 g
Catalog No. Quantity
01337857 125 g
01337856 2.5 kg
01337855 500 g
3 options

Catalog No. 01337857

Supplier: Labchem Inc C1281500125D2

Only null left

Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym Tripotassium Citrate Monohydrate
IUPAC Name tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
SMILES O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Boiling Point 230°C
Melting Point 180°C (anh)
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.98
Chemical Name or Material Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate)