missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate), Reagents

Catalog No. 01337857
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337857 125 g
01337856 2.5 kg
01337855 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337857 Supplier Labchem Inc Supplier No. C1281500125D2
Only null left

Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
Synonym Tripotassium Citrate Monohydrate
IUPAC Name tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
SMILES O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Boiling Point 230°C
Melting Point 180°C (anh)
Color Colorless/White
Physical Form Solid
Quantity 125 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.98
Chemical Name or Material Potassium Citrate Monohydrate, Reagent (Tripotassium Citrate Monohydrate)