missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate, Monohydrate, Granular, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18605532
Change view
Click to view available options
Quantity:
50 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605532 50 g Poly Bottle
18605529 500 g Poly Bottle
18605530 2.5 kg Poly Bottle
18605531 12 kg Poly Pail
18605533 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18605532 Supplier Spectrum Chemical Mfg Cor Supplier No. PO17550GM

PO175, 6100-05-6, C6H5K3O7·H2O

Spectrum™ Chemical Potassium Citrate, Monohydrate, Granular, USP belongs to a class of drugs known as urinary alkalinizers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6100-05-6
Molecular Formula C6H7K3O8
Molecular Weight (g/mol) 324.41
InChI Key PJAHUDTUZRZBKM-UHFFFAOYSA-K
IUPAC Name tripotassium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
SMILES O.[K+].[K+].[K+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Quantity 50 g
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 3-6%
Packaging Poly Bottle