missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate, Monohydrate, Crystal, 99%, Spectrum™ Chemical

Catalog No. 18612574
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612574 125 g Poly Bottle
18612575 500 g Poly Bottle
18612576 2.5 kg Poly Pail
18612577 12 kg Poly Pail
18612578 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612574 Supplier Spectrum Chemical Mfg Cor Supplier No. P1265125GM

Chemical Identifiers

CAS 6100-05-6

Specifications

pH 7.5 to 9.0
CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.