missing translation for 'onlineSavingsMsg'
Learn More

Potassium Citrate, Monobasic 98.0% (NT), MilliporeSigma™ Supelco™

Catalog No. p-7193404
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015707 250 g
111015708 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015707 Supplier Merck Emd Millipore Supplier No. 60215250G
Only null left

Chemical Identifiers

CAS 866-83-1
Molecular Formula C6H7KO7
Molecular Weight (g/mol) 230.21
MDL Number MFCD00036472
InChI Key WKZJASQVARUVAW-UHFFFAOYNA-M
Synonym Citric acid monopotassium salt; Potassium dihydrogen citrate
IUPAC Name potassium 3,4-dicarboxy-3-hydroxybutanoate
SMILES [K+].OC(=O)CC(O)(CC([O-])=O)C(O)=O

Specifications

Quantity 250 g
Linear Formula KH2C6H5O7
Formula Weight 230.21
Percent Purity ≥98.0% (NT)