missing translation for 'onlineSavingsMsg'
Learn More

Potassium, chunks, in mineral oil, 98%

Catalog No. p-7058010
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1326714 25 g
AAL1326722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1326714 Supplier Thermo Scientific Chemicals Supplier No. L1326714
Only null left

CAS: 7440-09-7 | K | 39.10 g/mol

Potassium forms a blue solution in tetrahydrofoliate which is used for the ? -monoalkylation of -lactones in the presence of 18-crown-6. It is used for generation of potassium ethoxide in solution, for use in the Birch reduction of an aromatic ring, for preparation of potassium amide,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium forms a blue solution in tetrahydrofoliate which is used for the ɑ -monoalkylation of -lactones in the presence of 18-crown-6. It is used for generation of potassium ethoxide in solution, for use in the Birch reduction of an aromatic ring, for preparation of potassium amide,

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 9-7-7440
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]

Specifications

Color Gray-Silver
Melting Point 63°C to 64°C
Boiling Point 765°C to 770°C
Density 0.860 g/mL
Quantity 25 g
Odor Odorless
Sensitivity Air and moisture sensitive
Form Solid
UN Number UN2257
Merck Index 14,7604
Solubility Information Reacts with water.
Formula Weight 39.1
Percent Purity 98%
Chemical Name or Material Potassium, chunks, in mineral oil
Show More Show Less
Hazard Category H260-H314
Hazard Statement GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P223-P231+P232-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: POTASSIUM
EINECSNumber 231-119-8
RTECSNumber TS6460000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only