missing translation for 'onlineSavingsMsg'
Learn More

Potassium chromate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Catalog No. p-7072010
Change view
Click to view available options
Quantity:
250 g
1 kg
Catalog No. Quantity
6001200 250 g
6001199 1 kg
2 options

Catalog No. 6001200

Supplier: Honeywell Chemicals 31250250G

Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7789-00-6
Molecular Formula CrK2O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00011368
InChI Key XMXNVYPJWBTAHN-UHFFFAOYSA-N
Synonym potassium chromate, bipotassium chromate, potassium chromate vi, dipotassium monochromate, dipotassium chromate, tarapacaite, chromate of potash, chromate of potass, neutral potassium chromate, caswell no. 687
PubChem CID 24597
ChEBI CHEBI:75249
SMILES [K+].[K+].[O-][Cr]([O-])(=O)=O

Specifications

Quantity 250 g
Linear Formula K2CrO4
UN Number 3288
IUPAC Name dipotassium;dioxido(dioxo)chromium
Formula Weight 194.19g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Density Vapor Density: 6.7 (vs air)
Chemical Name or Material Potassium Chromate, Reag. Ph. Eur.
Hazard Statement P201-P273-P280-P301 + P310 + P330-P304 + P340 + P312-P308 + P313
Precautionary Statement H301-H315-H317-H319-H335-H340-H350i-H410
EINECSNumber 232-140-5
RTECSNumber GB2940000