missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chromate, Granular, ACS, 99%, Spectrum™ Chemical
Specifications
Specifications
| pH | 8.6 to 9.8 |
| CAS Max % | 1 |
| Quantity | 25 g |
| Percent Purity | 99% |
| Grade | ACS |
| Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.