missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Solution, For Ag/AgCl electrodes, ∼3 M KCl, MilliporeSigma™ Supelco™

Catalog No. 111015420
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015420 1 L
111015418 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015420 Supplier Merck Emd Millipore Supplier No. 601371L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Saturated with silver chloride (∼0.16 M AgCl)

KCl exists as a white ionic solid, and can be prepared by neutralizing HCl with potassium hydroxide solution. It is less soluble in hot water as compared to cold water. It occurs naturally as the minerals carnallite and sylvine. It is generally used as a fertilizer.

Specifications

Chemical Name or Material Potassium Chloride
CAS 7447-40-7
Physical Form Liquid
Quantity 1 L
Linear Formula KCl
Synonym Potassium chloride solution saturated with AgCl
Molecular Weight (g/mol) 74.55
Formula Weight 74.55
Packaging Bottle
Density 1.13 g/mL (at 20°C)