missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Solution, For Ag/AgCl electrodes, ∼3 M KCl, MilliporeSigma™ Supelco™

Catalog No. 111015418
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015418 250 mL
111015420 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015418 Supplier Merck Emd Millipore Supplier No. 60137250ML

Saturated with silver chloride (∼0.16 M AgCl)

KCl exists as a white ionic solid, and can be prepared by neutralizing HCl with potassium hydroxide solution. It is less soluble in hot water as compared to cold water. It occurs naturally as the minerals carnallite and sylvine. It is generally used as a fertilizer.

Specifications

CAS 7447-40-7
Quantity 250 mL
Linear Formula KCl
Synonym Potassium chloride solution saturated with AgCl
Formula Weight 74.55
Density 1.13 g/mL (at 20°C)