missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Solution, BioUltra, ∼3 M in H2O, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 7447-40-7 |
| Quantity | 250 mL |
| Linear Formula | KCl |
| MDL Number | MFCD00011360 |
| Formula Weight | 74.55 |
| Density | 1.120 g/cm3 (at 20°C) |