missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, 4 Molar (Electrode Filling Solution) Saturated with Silver Chloride, Reagents

Catalog No. 01334505
Change view
Click to view available options
Quantity:
120 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334505 120 mL
01334504 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334505 Supplier Labchem Inc Supplier No. CP007400120A
Only null left

Potassium Chloride, 4 Molar (Electrode Filling Solution) Saturated with Silver Chloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 104°C
Chemical Name or Material Potassium Chloride, 4 Molar (Electrode Filling Solution) Saturated with Silver Chloride
CAS 7732-18-5, 7447-40-7
Color Colorless
pH 5.4 to 8.6
Physical Form Liquid
Quantity 120 mL
Molecular Formula KCl/H2O/AgCl
Synonym Muriate of Potash
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.17
Show More Show Less