missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, 3 Molar Electrode Filling Solution Saturated with Silver Chloride, Reagents

Catalog No. 01334503
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01334503 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334503 Supplier Labchem Inc Supplier No. CP007500500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Chloride, 3 Molar Electrode Filling Solution Saturated with Silver Chloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Chloride, 3 Molar Electrode Filling Solution Saturated with Silver Chloride
Boiling Point 103°C
Color Colorless
CAS 7732-18-5, 7447-40-7
pH 5.4 to 8.6
Physical Form Liquid
Quantity 500 mL
Molecular Formula KCl/H2O/AgCl
Synonym Muriate of Potash
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.1111
Show More Show Less