missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chlorate, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99%
$3550.88 - $3550.88
Chemical Identifiers
| CAS | 3811-04-9 |
|---|---|
| Molecular Formula | ClKO3 |
| Molecular Weight (g/mol) | 122.55 |
| MDL Number | MFCD00011361 |
| InChI Key | VKJKEPKFPUWCAS-UHFFFAOYSA-M |
| Synonym | potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt |
| PubChem CID | 6426889 |
| IUPAC Name | potassium;chlorate |
| SMILES | [O-]Cl(=O)=O.[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6001197
|
Honeywell-Fluka
3124725KG |
25 kg |
N/A
|
N/A | |||||
|
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. |
|||||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 3811-04-9 | |
| 122.55 | |
| VKJKEPKFPUWCAS-UHFFFAOYSA-M | |
| 6426889 | |
| [O-]Cl(=O)=O.[K+] |
| ClKO3 | |
| MFCD00011361 | |
| potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt | |
| potassium;chlorate |
Specifications
| 3811-04-9 | |
| ClKO3 | |
| MFCD00011361 | |
| potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt | |
| [O-]Cl(=O)=O.[K+] | |
| 122.55 | |
| 122.55g/mol | |
| ACS/puriss. p.a. |
| 25 kg | |
| KClO3 | |
| 1485 | |
| VKJKEPKFPUWCAS-UHFFFAOYSA-M | |
| potassium;chlorate | |
| 6426889 | |
| ≥99% | |
| Potassium Chlorate, Reag. Ph. Eur. |
Safety and Handling
P220-P273
H271-H302 + H332-H411
EINECSNumber : 223-289-7
RTECSNumber : FO0350000