missing translation for 'onlineSavingsMsg'
Learn More

Potassium Butylxanthate 95.0+%, TCI America™

Catalog No. B074725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B074725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B074725G Supplier TCI America Supplier No. B074725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 871-58-9
Molecular Formula C5H9KOS2
Molecular Weight (g/mol) 188.344
MDL Number MFCD00059688
InChI Key OMKVZYFAGQKILB-UHFFFAOYSA-M
Synonym Butylxanthic Acid Potassium Salt
PubChem CID 11424037
IUPAC Name potassium;butoxymethanedithioate
SMILES CCCCOC(=S)[S-].[K+]

Specifications

Quantity 25 g
Formula Weight 188.34
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Butylxanthate
EINECSNumber (2)-1787
RTECSNumber FG1223000
TSCA No
Recommended Storage Refrigerator