missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromomethyltrifluoroborate, 95%, Thermo Scientific Chemicals

Catalog No. AAH3216806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3216806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3216806 Supplier Thermo Scientific Chemicals Supplier No. H3216806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 888711-44-2 | Molecular Formula: CH2BBrF3K | Molecular Weight: 200.835 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 888711-44-2
Molecular Formula CH2BBrF3K
Molecular Weight (g/mol) 200.835
MDL Number MFCD09265154
InChI Key AZDFPIRYUOCVCJ-UHFFFAOYSA-N
Synonym potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v
PubChem CID 23690312
IUPAC Name potassium;bromomethyl(trifluoro)boranuide
SMILES [B-](CBr)(F)(F)F.[K+]

Specifications

Linear Formula BrCH2BF3K
Quantity 5 g
Formula Weight 200.84
Percent Purity 95%
Chemical Name or Material Potassium bromomethyltrifluoroborate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only