missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium (Bromomethyl)trifluoroborate 98.0+%, TCI America™
$61.61 - $246.43
Chemical Identifiers
| CAS | 888711-44-2 |
|---|---|
| Molecular Formula | CH2BBrF3K |
| Molecular Weight (g/mol) | 200.835 |
| MDL Number | MFCD09265154 |
| InChI Key | AZDFPIRYUOCVCJ-UHFFFAOYSA-N |
| Synonym | potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v |
| PubChem CID | 23690312 |
| IUPAC Name | potassium;bromomethyl(trifluoro)boranuide |
| SMILES | [B-](CBr)(F)(F)F.[K+] |
Chemical Identifiers
| 888711-44-2 | |
| 200.835 | |
| AZDFPIRYUOCVCJ-UHFFFAOYSA-N | |
| 23690312 | |
| [B-](CBr)(F)(F)F.[K+] |
| CH2BBrF3K | |
| MFCD09265154 | |
| potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v | |
| potassium;bromomethyl(trifluoro)boranuide |
Specifications
| 888711-44-2 | |
| White-Yellow | |
| MFCD09265154 | |
| potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v | |
| [B-](CBr)(F)(F)F.[K+] | |
| 200.835 | |
| 200.83 | |
| Crystalline Powder |
| 220°C | |
| CH2BBrF3K | |
| 1 g | |
| AZDFPIRYUOCVCJ-UHFFFAOYSA-N | |
| potassium;bromomethyl(trifluoro)boranuide | |
| 23690312 | |
| ≥98.0% (T) | |
| Potassium (Bromomethyl)trifluoroborate |
Safety and Handling
TSCA : No