missing translation for 'onlineSavingsMsg'
Learn More

Potassium (Bromomethyl)trifluoroborate 98.0+%, TCI America™

Catalog No. P18075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18075G 5 g
P18071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18075G Supplier TCI America Supplier No. P18075G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 888711-44-2
Molecular Formula CH2BBrF3K
Molecular Weight (g/mol) 200.835
MDL Number MFCD09265154
InChI Key AZDFPIRYUOCVCJ-UHFFFAOYSA-N
Synonym potassium bromomethyl trifluoroborate, potassium bromomethyltrifluoroborate, potassium bromomethyl trifluoroboranuide, potassium bromomethyl trifluoro boranuide, potassium bromomethyl trifluoro borate 1-, borate 1-, bromomethyl trifluoro-, potassium 1:1 , t-4, pubchem11564, bromomethyl potassium trifluoroborate, postassium bromomethyltrifluoroborate, trifluoro bromomethyl potassioboron v
PubChem CID 23690312
IUPAC Name potassium;bromomethyl(trifluoro)boranuide
SMILES [B-](CBr)(F)(F)F.[K+]

Specifications

Melting Point 220°C
Color White-Yellow
Quantity 5 g
Formula Weight 200.83
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium (Bromomethyl)trifluoroborate