Learn More
Potassium bromide, for spectroscopy, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 457320025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Potassium bromide | |
| 2.5kg | |
| KBr | |
| Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol | |
| [K+].[Br-] | |
| 119 | |
| CHEBI:32030 | |
| ≥99.0% (Argentometry) | |
| Authentic |
| 7758-02-3 | |
| BrK | |
| potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095 | |
| IOLCXVTUBQKXJR-UHFFFAOYSA-M | |
| potassium;bromide | |
| 253877 | |
| 119 | |
| Spectroscopy |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 231-830-3
RUO â Research Use Only