missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, Spectroscopy Grade

Catalog No. p-7050940
Change view
Click to view available options
Quantity:
0.25 lb.
1 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39796Q4 0.25 lb.
AA39796P2 1 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39796Q4 Supplier Thermo Scientific Chemicals Supplier No. 039796Q4
Only null left

CAS: 7758-02-3 | BrK | 119.00 g/mol

Manufacturing of photographic papers, process engraving and lithography, spectroscopy, as an analytical lab reagent. Potassium bromide is applied as a veterinary drug and antiepileptic medication for dogs. It is an utilitarian for the bromination of alkenes using cerium(IV) ammonium nitrate. It acts as an essential role with diacetoxy iodobenzene for the efficient

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Color White
Physical Form Random Cuttings, Chunks
Quantity 0.25 lb.
Merck Index 14,7618
Solubility Information Soluble in water and very slightly soluble in diethyl ether.
Formula Weight 119.01
Grade Spectroscopy
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Density 2.75 g/mL
Chemical Name or Material Potassium bromide
Show More Show Less
Hazard Category H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 231-830-3
RTECSNumber TS7650000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only