missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, puriss., Honeywell Fluka™

Catalog No. p-7071695
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Catalog No. Quantity
6001795 100 g
6001799 500 g
6001796 1 kg
6001797 2.5 kg
60044466 6 x 1 kg
60044467 6 x 2.5 kg
6 options

Catalog No. 6001795

Supplier: Honeywell Chemicals 02110100G

Only null left

Puriss., meets analytical specification of BP, Ph. Eur., USP, 99.5-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
SMILES [K+].[Br-]

Specifications

CAS 7758-02-3, 14708-99-7
Quantity 100 g
Linear Formula KBr
UN Number NONH for all modes of transport
IUPAC Name potassium bromide
Formula Weight 119g/mol
Percent Purity 99.5 to 100.5%
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 0.01mmHg (20°C), 1mmHg (795°C)
Chemical Name or Material Potassium Bromide, Meets analytical specification of Ph. Eur.BP,USP
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P337 + P313
Precautionary Statement H319
EINECSNumber 231-830-3
RTECSNumber TS7650000