missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, FT-IR Grade, ≥99% trace metals basis, Honeywell Fluka™

Catalog No. p-7122136
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60045328 25 g
6002131 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60045328 Supplier Honeywell Chemicals Supplier No. 22186425G
Only null left

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1,383°C
Melting Point 734°C (literature)
CAS Max % 100.0000%
Quantity 25 g
Linear Formula KBr
UN Number NONH for all modes of transport
IUPAC Name potassium bromide
Formula Weight 119
Percent Purity >99%
Packaging Poly Bottle
Vapor Pressure 1mmHg (795°C), 0.01mmHg (20°C)
Density 2.73 g/cm3
Chemical Name or Material Potassium bromide, FT-IR Grade, ≥99% trace metals basis
Show More Show Less
Hazard Statement P280-P305 + P351 + P338-P337 + P313
Precautionary Statement H319
ShelfLife 1080 days from date of manufacture
EINECSNumber 231-830-3
RTECSNumber TS7650000
Recommended Storage Room Temp