missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, MilliporeSigma™

Catalog No. M1049070100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1049070100 100 g Glass Bottle
M1049070500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1049070100 Supplier Technidata Supplier No. 1.04907.0100

For IR spectroscopy Uvasol™

  • Potassium bromide is transparent from near UV to long wave IR wavelengths.
  • In IR-spectroscopy, solid samples which are difficult to melt or dissolve in any suitable IR-transmitting solvent, are analyzed by grinding with Potassium bromide powder, and pressing into disc.
  • Potassium bromide Uvasol™ is adjusted to a mean particle size of 150 μm.
  • Sufficient for preparation of perfectly good pellets without need for further pre-treatment and associated risk of contamination.
  • It also retains its powdery form over a period of years if stored in an air-tight condition.
  • Physical suitability for pelletising is checked by special application test and its chemical purity established by full spectrum FT-IR analysis.

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
pH 5.5 to 8.5 (50g/L, H2O, 20°C)
Quantity 100 g
Solubility Information Soluble in water: 650g/L at 10°C
Formula Weight 119
Odor Odorless
Packaging Glass Bottle
Vapor Pressure 1.3hPa at 795°C
Density 2.75g/cm3 at 20°C
Chemical Name or Material Potassium bromide
Show More Show Less
Hazard Category Irritant
EINECSNumber 231-830-3
RTECSNumber TS7650000
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry.