missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide, Crystal, IR Grade, Spectrum™ Chemical
P1225, 7758-02-3, KBr
$460.43 - $2272.23
Chemical Identifiers
| CAS | 2-3-7758 |
|---|---|
| Molecular Formula | BrK |
| Molecular Weight (g/mol) | 119.00 |
| MDL Number | MFCD00011358 |
| InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
| IUPAC Name | potassium bromide |
| SMILES | [K+].[Br-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605473
|
Spectrum Chemical Mfg Cor
P122525GM |
25 g | Amber Glass Bottle |
Each for $460.43
|
|
||||
|
18605475
|
Spectrum Chemical Mfg Cor
P1225100GM |
100 g | Amber Glass Bottle |
Each for $1,418.77
|
|
||||
|
18605474
|
Spectrum Chemical Mfg Cor
P122525GMCS6 |
6 x 25 g | Amber Glass Bottle |
Case of 6 Each for $2,272.23
|
|
||||