missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, BAKER INSTRA-ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5390812
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003719 25 g Wide Mouth Amber Glass Bottle
02003718 100 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003719 Supplier Avantor Supplier No. 296103
Only null left

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    For Infrared Analysis

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Quantity 25 g
IUPAC Name potassium bromide
Formula Weight 118.99
Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.75kg
Chemical Name or Material Potassium Bromide