missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide, ACS Grade, ≥ 99.0%, LabChem™
Specifications
Specifications
| Boiling Point | 1,435°C |
| Melting Point | 730°C |
| Color | White |
| CAS Max % | 100 |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Linear Formula | KBr |
| Solubility Information | Soluble in water |
| Formula Weight | 119 |
| Percent Purity | ≥99.0% |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation and serious eye irritation. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Avoid breathing dust. Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. Use only outdoors or in a well-ventilated area. Avoid release to the environment. If on skin: Wash with plenty of water and soap. Take off contaminated clothing and wash before reuse. If skin irritation occurs: Get medical advice/attention. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. Store locked up in a well-ventilated place. Keep container tightly closed. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |