missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, ACS, 99% min

Catalog No. p-4407720
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1261218 50 g
AA1261230 250 g
AA12612A1 1 kg
AA12612A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1261218 Supplier Thermo Scientific Chemicals Supplier No. 01261218
Only null left

For Manufacturing of Photographic Papers, Process Engraving and Lithography, Spectroscopy | CAS: 7758-02-3 | BrK | 119.00 g/mol

Used in manufacturing of photographic papers, engraving and lithography, in spectroscopy, and as an analytical lab reagent. Potassium bromide is applied as a veterinary drug, as an antiepileptic medication for dogs. It is utilized for the bromination of alkenes using cerium(IV) ammonium nitrate. It plays an essential role with diacetoxy iodobenzene for the efficient bromination of alkenes and alkynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
SMILES [K+].[Br-]

Specifications

Boiling Point 1435°C
Melting Point 730°C
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99% min.
Solubility Information Freely soluble in water. Soluble in alcohol,glycerol.
IUPAC Name potassium bromide
Formula Weight 119.01
Grade ACS
Odor Odorless
Refractive Index 1.559
Sensitivity Hygroscopic
Appearance White powder/crystalline/beads
Density 2.75 g/cm3 at 20°C
Chemical Name or Material Potassium bromide, For analysis ACS
Show More Show Less
Hazard Category H319-H335
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 231-830-3
RTECSNumber TS7650000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only