missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, 99+%

CAS: 7758-02-3 | BrK | 119.00 g/mol

Supplier:  Thermo Scientific Chemicals A163390B

 View more versions of this product

Catalog No. AAA163390B


Only null left
Explore available promotions

Description

Description

Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

Solubility
Slightly soluble in alcohol but insoluble in acetone and ethanol.

Notes
Strong oxidizer. Incompatible with combustible materials, organics, reducing agents, aluminum and finely powdered metals
Specifications

Specifications

1435°C
730°C
1000 g
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium;bromide
253877
119.01
Odorless
Hygroscopic
Potassium bromide
2-3-7758
BrK
14,7618
Slightly soluble in alcohol but insoluble in acetone and ethanol.
[K+].[Br-]
119.00
CHEBI:32030
≥99%
1.559
2.75

Chemical Identifiers

2-3-7758
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
[K+].[Br-]
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c

H319-H335

EINECSNumber : 231-830-3

RTECSNumber : TS7650000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only