missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium bromide, 99+%, for biochemistry

CAS: 7758-02-3 | BrK | 119.00 g/mol

Supplier:  Thermo Scientific Chemicals 212580010

 View more versions of this product

Catalog No. AC212580010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For biochemistry
Specifications

Specifications

1435.0°C
734.0°C
1 kg
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
253877
119
Biochemical
Potassium bromide
2-3-7758
99% min. (Argentometry)
KBr
15,7739
Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol
[K+].[Br-]
119.00
CHEBI:32030
≥99%
Glass bottle

Chemical Identifiers

2-3-7758
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium bromide
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
[K+].[Br-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 231-830-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only