missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromide, 99.999%, (trace metal basis)

Catalog No. AC436950100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436950100 10 g Plastic bottle
AC436950500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436950100 Supplier Thermo Scientific Chemicals Supplier No. 436950100

CAS: 7758-02-3 | BrK | 119.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
Synonym potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
PubChem CID 253877
ChEBI CHEBI:32030
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

Boiling Point 1435.0°C
Melting Point 734.0°C
Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 33% (K) Typical (Argentometry)
Linear Formula KBr
Merck Index 15,7739
Solubility Information Solubility in water: 650g/L (20°C). Other solubilities: 1020g/L water (100°C), 1.42g/L ethanol (25°C), 217g/L glycerol, 4g/L alcohol
Formula Weight 119
Percent Purity 99.999%
Grade Trace Metal Basis
Packaging Plastic bottle
Chemical Name or Material Potassium bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 231-830-3

RUO – Research Use Only