missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate Solution, Volumetric, 1/60 M KBrO3 (0.1N), Solstice

Catalog No. 60046328
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046328 1 L Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60046328 Supplier Solstice Supplier No. 342681L

Volumetric, 1/60 M KBrO3 (0.1N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Physical Form Liquid
Quantity 1 L
Linear Formula KBrO3
UN Number NONH for all modes of transport
Formula Weight 167g/mol
Packaging Poly bottle
Chemical Name or Material Potassium bromate solution
Hazard Statement P201-P308 + P313