missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromate, Granular, ACS, 99.8%, Spectrum™ Chemical

Catalog No. p-7166041
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612492 25 g Poly Bottle
18612494 500 g Poly Bottle
18612495 2.5 kg Poly Pail
18612496 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612492 Supplier Spectrum Chemical Mfg Cor Supplier No. P121525GM
Only null left

Chemical Identifiers

CAS 1-2-7758

Specifications

pH 5.0to 9.0
CAS Max % 1
Quantity 25 g
Percent Purity 99.8%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.