missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate concentrate, For 1L standard solution, 1/60 M KBrO3 (0.1N), Honeywell Fluka™

Catalog No. p-7122103
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002666 1 Pc.
60046690 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002666 Supplier Honeywell Chemicals Supplier No. 380801EA
Only null left

for 1L standard solution, 1/60 M KBrO3 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

CAS 1-2-7758
Quantity 1 Pc.
Linear Formula KBrO3
UN Number NONH for all modes of transport
Formula Weight 167g/mol
Packaging Ampoule
Chemical Name or Material Potassium bromate concentrate
Hazard Statement P201-P308 + P313