missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate concentrate, For 1L standard solution, 1/60 M KBrO3 (0.1N), Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| CAS | 1-2-7758 |
| Physical Form | Liquid |
| Quantity | 1 Pc. |
| Linear Formula | KBrO3 |
| UN Number | NONH for all modes of transport |
| Formula Weight | 167g/mol |
| Grade | Not Available |
| Packaging | Ampoule |
| Chemical Name or Material | Potassium bromate concentrate |
Safety and Handling
| Hazard Statement | P201-P308 + P313 |