missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bromate, ACS, 99.8% min
Description
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 370°C (decomposition) |
| Physical Form | Granules |
| Quantity | 500 g |
| Assay Percent Range | 99.8% min |
| Density | 3.27 g/mL |
| Solubility Information | Slightly soluble in alcohol but insoluble in acetone and ethanol. |
| Formula Weight | 167 |
| Grade | ACS Reagent |
| Chemical Name or Material | Potassium bromate |
Safety and Handling
| Hazard Category | H271-H301-H335-H336-H351-H373 |
| Precautionary Statement | P201-P202-P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P310-P304+P340-P306+P360-P308+P313-P330-P370+P378q-P371+P380+P375-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: II |
| EINECSNumber | 231-829-8 |
| RTECSNumber | EF8725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only