missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate, ACS, 99.8% min

Catalog No. AA4001322
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4001322 100 g
AA4001336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4001322 Supplier Thermo Scientific Chemicals Supplier No. 04001322

CAS: 7758-01-2 | BrKO3 | 167.00 g/mol

Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Melting Point 370°C (decomposition)
Physical Form Granules
Quantity 100 g
Assay Percent Range 99.8% min
Solubility Information Slightly soluble in alcohol but insoluble in acetone and ethanol.
Formula Weight 167
Grade ACS Reagent
Density 3.27 g/mL
Chemical Name or Material Potassium bromate
Hazard Category H271-H301-H335-H336-H351-H373
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P310-P304+P340-P306+P360-P308+P313-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 231-829-8
RTECSNumber EF8725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only