missing translation for 'onlineSavingsMsg'
Learn More

Potassium bromate, 99%

Catalog No. aaa1825822
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1825822 100 g
AAA1825836 500 g
AAA182580E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1825822 Supplier Thermo Scientific Chemicals Supplier No. A1825822

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7758-01-2 | BrKO3 | 167.00 g/mol

Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an analytical reagent and an oxidizing agent. Potassium bromate is used primarily as a maturing agent for flour and as a dough conditioner. It is also used as a laboratory reagent, in permanent-wave compounds such as food additives and explosives.

Solubility
Slightly soluble in alcohol but insoluble in acetone and ethanol.

Notes
Strong oxidizer. Incompatible with combustible materials, organics, reducing agents, aluminum and finely powdered metals

Chemical Identifiers

CAS 1-2-7758
Molecular Formula BrKO3
Molecular Weight (g/mol) 167.00
MDL Number MFCD00011359
InChI Key OCATYIAKPYKMPG-UHFFFAOYSA-M
Synonym potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
PubChem CID 23673461
ChEBI CHEBI:38211
IUPAC Name potassium;bromate
SMILES [O-]Br(=O)=O.[K+]

Specifications

Melting Point ∼370°C (decomposition)
Quantity 100 g
Odor Odorless
Density 3.27
UN Number UN1484
Merck Index 14,7617
Solubility Information Slightly soluble in alcohol but insoluble in acetone and ethanol.
Formula Weight 167
Percent Purity 99%
Chemical Name or Material Potassium bromate
Hazard Category H271-H301-H335-H336-H351-H373
Hazard Statement GHS H Statement
H271-H301
May cause fire or explosion; strong oxidizer.
Toxic if swallowed.
Precautionary Statement P201-P202-P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P310-P304+P340-P306+P360-P308+P313-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM BROMATE
EINECSNumber 231-829-8
RTECSNumber EF8725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only