missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bisnonafluoro-1-butanesulfonimidate 98.0+%, TCI America™

Catalog No. p-7133949
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07121G 1 g
N07125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07121G Supplier TCI America Supplier No. N07121G
Only null left

Chemical Identifiers

CAS 129135-87-1
Molecular Formula C8F18KNO4S2
Molecular Weight (g/mol) 619.281
MDL Number MFCD06200794
InChI Key XESFGOBWYAMCMB-UHFFFAOYSA-N
Synonym Bisnonafluoro-1-butanesulfonimide Potassium Salt, Bis(nonafluoro-1-butanesulfonyl)amine Potassium Salt, Nonafluoro-1-butanesulfonimide Potassium Salt, Potassium Nonafluoro-1-butanesulfonimidate
PubChem CID 11319533
IUPAC Name potassium;bis(1,1,2,2,3,3,4,4,4-nonafluorobutylsulfonyl)azanide
SMILES C(C(C(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(C(F)(F)F)(F)F.[K+]

Specifications

Color White
Quantity 1 g
Formula Weight 619.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Bisnonafluoro-1-butanesulfonimidate