missing translation for 'onlineSavingsMsg'
Learn More

Potassium bis(trifluoromethylsulfonyl)imide

Catalog No. p-7080247
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46909MD 250 mg
AA4690903 1 g
AA4690906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA46909MD Supplier Thermo Scientific Chemicals Supplier No. 046909MD
Only null left

CAS: 90076-67-8 | C2F6KNO4S2 | 319.234 g/mol

Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90076-67-8
Molecular Formula C2F6KNO4S2
Molecular Weight (g/mol) 319.234
MDL Number MFCD06200829
InChI Key KVFIZLDWRFTUEM-UHFFFAOYSA-N
Synonym potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide
PubChem CID 11099217
IUPAC Name potassium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+]

Specifications

Melting Point 198°C to 203°C
Linear Formula (CF3SO2)2NK
Quantity 250 mg
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information It is soluble in water. 10mg/mL.
Formula Weight 319.24
Physical Form Solid
Chemical Name or Material Potassium bis (trifluoromethylsulfonyl)imide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only