missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bis(fluorosulfonyl)imide 95.0+%, TCI America™
Supplier: TCI America P250925G
Specifications
| Potassium Bis(fluorosulfonyl)imide | |
| 101°C | |
| F2KNO4S2 | |
| 25 g | |
| MHEBVKPOSBNNAC-UHFFFAOYSA-N | |
| potassium;bis(fluorosulfonyl)azanide | |
| 23697682 | |
| ≥95.0% (T) |
| 14984-76-0 | |
| White | |
| MFCD17018950 | |
| 1759 | |
| [N-](S(=O)(=O)F)S(=O)(=O)F.[K+] | |
| 219.218 | |
| 219.22 | |
| Crystalline Powder |
Chemical Identifiers
| 14984-76-0 | |
| 219.218 | |
| MHEBVKPOSBNNAC-UHFFFAOYSA-N | |
| potassium;bis(fluorosulfonyl)azanide |
| F2KNO4S2 | |
| MFCD17018950 | |
| 23697682 | |
| [N-](S(=O)(=O)F)S(=O)(=O)F.[K+] |
Safety and Handling
TSCA : No