missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bis(fluorosulfonyl)imide 95.0+%, TCI America™

Catalog No. P25095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25095G 5 g
P250925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25095G Supplier TCI America Supplier No. P25095G
Only null left

Chemical Identifiers

CAS 14984-76-0
Molecular Formula F2KNO4S2
Molecular Weight (g/mol) 219.218
MDL Number MFCD17018950
InChI Key MHEBVKPOSBNNAC-UHFFFAOYSA-N
PubChem CID 23697682
IUPAC Name potassium;bis(fluorosulfonyl)azanide
SMILES [N-](S(=O)(=O)F)S(=O)(=O)F.[K+]

Specifications

Melting Point 101°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 219.22
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Bis(fluorosulfonyl)imide