missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bis(1-pyrazolyl)borohydride 98.0+%, TCI America™

Catalog No. p-7135137
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14391G 1 g
P14395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14391G Supplier TCI America Supplier No. P14391G
Only null left

Chemical Identifiers

CAS 18583-59-0
Molecular Formula C6H6BKN4
Molecular Weight (g/mol) 184.05
MDL Number MFCD00058739
InChI Key KIXPNQZZOUCWGN-UHFFFAOYSA-N
Synonym Potassium Dihydrobis(1-pyrazolyl)borate
PubChem CID 44630456
IUPAC Name potassium;di(pyrazol-1-yl)boranide
SMILES [B-](N1C=CC=N1)N2C=CC=N2.[K+]

Specifications

Melting Point 168°C
Color White
Quantity 1 g
UN Number 2813
Formula Weight 186.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium Bis(1-pyrazolyl)borohydride