missing translation for 'onlineSavingsMsg'
Learn More

Potassium Biphthalate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003711
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003711 100 g Poly Bottle
02003713 500 g Poly Bottle
02003710 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02003711 Supplier Avantor Supplier No. 295800

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Physical Form Crystals
Quantity 100 g
Formula Weight 204.22
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.64kg
Chemical Name or Material Potassium Biphthalate, Acidimetric Standard